missing translation for 'onlineSavingsMsg'
Learn More

(2S)-2-Amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic Acid (Diiodothyronine), Mikromol™

Product Code. 17949214
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17949214 25 mg 25mg
17998244 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17949214 Supplier Mikromol Supplier No. MM0626.080025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S)-2-Amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic Acid (Diiodothyronine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S)-2-Amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic Acid (3,5-Diiodo-L-thyronine)
CAS 1041-01-6
Analyte or Component Names (2S)-2-Amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic Acid (3,5-Diiodo-L-thyronine)
Shipping Condition Room Temperature
Molecular Formula C15 H13 I2 N O4
Certifications/Compliance ISO 9001
Synonym Liothyronine Sodium Imp. E (EP), Liothyronine Imp. E (EP), L-Tyrosine, O-(4-hydroxyphenyl)-3,5-diiodo-, Alanine, 3-[4-(p-hydroxyphenoxy)-3,5-diiodophenyl]-, L- (8CI), 3,5-Diiodo-L-thyronine, 3,5-Diiodothyronine, Diiodo-L-thyronine, Diiodothyronine, L-3,5-Diiodothyronine, L-Diiodothyronine, L-T2, NSC 90469, O-(4-Hydroxyphenyl)-3,5-diiodo-L-tyrosine, Levothyroxine Sodium Impurity E, Liothyronine Sodium Impurity E, Levothyroxine Impurity E, Liothyronine Impurity E
InChI Formula InChI=1S/C15H13I2NO4/c16-11-5-8(7-13(18)15(20)21)6-12(17)14(11)22-10-3-1-9(19)2-4-10/h1-6,13,19H,7,18H2,(H,20,21)/t13-/m0/s1
SMILES N[C@@H](Cc1cc(I)c(Oc2ccc(O)cc2)c(I)c1)C(=O)O
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic acid
Molecular Weight (g/mol) 525.08
Quantity 25 mg
Formula Weight 524.8934 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.