missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS)-2-[(Dimethylamino)methyl]cyclohexanone Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(2RS)-2-[(Dimethylamino)methyl]cyclohexanone Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-[(Dimethylamino)methyl]cyclohexanone Hydrochloride |
| CAS | 42036-65-7 |
| Analyte or Component Names | (2RS)-2-[(Dimethylamino)methyl]cyclohexanone Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H17 N O . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Tramadol Hydrochloride Imp. E (EP), Tramadol Imp. E (EP), Tramadol USP Related Compound B, Tramadol USP RC B, (2RS)-2-[(Dimethylamino)methyl]cyclohexanone Hydrochloride, Tramadol Hydrochloride Imp. E (EP) as Hydrochloride, Tramadol Related Compound B as Hydrochloride, Tramadol Hydrochloride Impurity E as Hydrochloride, Tramadol Impurity E as Hydrochloride |
| InChI Formula | InChI=1S/C9H17NO.ClH/c1-10(2)7-8-5-3-4-6-9(8)11;/h8H,3-7H2,1-2H3;1H |
| SMILES | Cl.CN(C)CC1CCCCC1=O |
| IUPAC Name | 2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction