missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-2-(Biphenyl-4-yl)propanoic Acid, Mikromol™

Product Code. 17943876
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943876 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17943876 Supplier Mikromol Supplier No. MM0727.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-2-(Biphenyl-4-yl)propanoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-2-(biphenyl-4-yl)propanoic acid
CAS 6341-72-6
Analyte or Component Names (2RS)-2-(biphenyl-4-yl)propanoic acid
Shipping Condition Room Temperature
Molecular Formula C15 H14 O2
Certifications/Compliance ISO 17025
Synonym Flurbiprofen Imp. A (EP), Flurbiprofen USP Related Compound A, Flurbiprofen USP RC A, (2RS)-2-(Biphenyl-4-yl)propanoic Acid, Flurbiprofen Related Compound A (USP), 2-(Biphenyl-4-yl)propionic acid, Biprofen, 2-(4-Biphenylyl)propionic Acid, Flurbiprofen Related Compound A, Flurbiprofen Impurity A
InChI Formula InChI=1S/C15H14O2/c1-11(15(16)17)12-7-9-14(10-8-12)13-5-3-2-4-6-13/h2-11H,1H3,(H,16,17)
SMILES CC(C(=O)O)c1ccc(cc1)c2ccccc2
IUPAC Name 2-(4-phenylphenyl)propanoic acid
Molecular Weight (g/mol) 226.27
Quantity 50 mg
Formula Weight 226.0994 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.