missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-2-Amino-4-phenylbutanoic Acid, Mikromol™

Product Code. 17989474
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17989474 25 mg 25mg
17989104 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17989474 Supplier Mikromol Supplier No. MM0598.010025
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-2-Amino-4-phenylbutanoic Acid
TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material (2RS)-2-Amino-4-phenylbutanoic Acid
CAS 1012-05-1
Analyte or Component Names (2RS)-2-Amino-4-phenylbutanoic Acid
Shipping Condition Room Temperature
Molecular Formula C10 H13 N O2
Certifications/Compliance ISO 17025
Synonym Lisinopril Dihydrate Imp. A (EP), Lisinopril Imp. A (EP), (2RS)-2-Amino-4-phenylbutanoic Acid, Lisinopril Dihydrate Impurity A, Lisinopril Impurity A
InChI Formula InChI=1S/C10H13NO2/c11-9(10(12)13)7-6-8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)
SMILES NC(CCc1ccccc1)C(=O)O
IUPAC Name 2-amino-4-phenylbutanoic acid
Molecular Weight (g/mol) 179.22
Quantity 25 mg
Formula Weight 179.0946 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.