missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-1-(Ethylamino)-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol, Mikromol™

Product Code. 17901676
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17901676 100 mg 100mg
17900456 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17901676 Supplier Mikromol Supplier No. MM0027.04
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-1-(Ethylamino)-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-1-(Ethylamino)-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol
CAS 109632-08-8
Analyte or Component Names (2RS)-1-(Ethylamino)-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol
Shipping Condition Room Temperature
Molecular Formula C14 H23 N O3
Certifications/Compliance ISO 17025
Synonym Metoprolol Succinate Imp. A (EP), Metoprolol Tartrate Imp. A (EP), Metoprolol Imp. A (EP), Metoprolol USP Related Compound A, Metoprolol USP RC A, (2RS)-1-(Ethylamino)-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol, Metoprolol Related Compound A, Metoprolol Succinate Impurity A, Metoprolol Tartrate Impurity A, Metoprolol Impurity A, Metoprolol Impurity A
InChI Formula InChI=1S/C14H23NO3/c1-3-15-10-13(16)11-18-14-6-4-12(5-7-14)8-9-17-2/h4-7,13,15-16H,3,8-11H2,1-2H3
SMILES CCNCC(O)COc1ccc(CCOC)cc1
IUPAC Name 1-(ethylamino)-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol
Molecular Weight (g/mol) 253.34
Quantity 100 mg
Formula Weight 253.1678 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.