missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-1-[4-(2-Hydroxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol, Mikromol™

Product Code. 30051927
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051927 25 mg 25mg
30052507 100 mg 100mg
30457640 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30051927 Supplier Mikromol Supplier No. MM0027.100025
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-1-[4-(2-Hydroxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-1-[4-(2-Hydroxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol
CAS 62572-94-5
Analyte or Component Names (2RS)-1-[4-(2-Hydroxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol
Shipping Condition Room Temperature
Molecular Formula C14 H23 N O3
Certifications/Compliance ISO 17025
Synonym Betaxolol Hydrochloride Imp. B (EP), Betaxolol Imp. B (EP), Metoprolol Succinate Imp. H (EP), Metoprolol Tartrate Imp. H (EP), Metoprolol Imp. H (EP), (2RS)-1-[4-(2-Hydroxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol, Betaxolol Hydrochloride Impurity B, Metoprolol Succinate Impurity H, Metoprolol Tartrate Impurity H, Betaxolol Impurity B, Metoprolol Impurity H
InChI Formula InChI=1S/C14H23NO3/c1-11(2)15-9-13(17)10-18-14-5-3-12(4-6-14)7-8-16/h3-6,11,13,15-17H,7-10H2,1-2H3
SMILES CC(C)NCC(O)COc1ccc(CCO)cc1
IUPAC Name 1-[4-(2-hydroxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
Molecular Weight (g/mol) 253.34
Quantity 25 mg
Formula Weight 253.1678 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.