missing translation for 'onlineSavingsMsg'
Learn More

(2R)-Vildagliptin, TRC

Product Code. 30011493
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011493 10 mg 10mg
30003218 250 mg 250mg
30010174 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011493 Supplier Toronto Research Chemicals Supplier No. TRCV30500510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1036959-27-9
Molecular Formula C17H25N3O2
Molecular Weight (g/mol) 303.4
Synonym 2-Pyrrolidinecarbonitrile, 1-[2-[(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)amino]acetyl]-, (2R)-, (2R)-1-[2-[(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)amino]acetyl]-2-pyrrolidinecarbonitrile, (R)-Vildagliptin
IUPAC Name (2R)-1-[2-[(3-hydroxy-1-adamantyl)amino]acetyl]pyrrolidine-2-carbonitrile
SMILES OC12CC3CC(C1)CC(C3)(C2)NCC(=O)N4CCC[C@@H]4C#N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C17H25N3O2/c18-9-14-2-1-3-20(14)15(21)10-19-16-5-12-4-13(6-16)8-17(22,7-12)11-16/h12-14,19,22H,1-8,10-11H2/t12?,13?,14-,16?,17?/m1/s1
Formula Weight 303.1947
Percent Purity >95%
Chemical Name or Material (R)-Vildagliptin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.