missing translation for 'onlineSavingsMsg'
Learn More

(2R-trans)-5,5-Dimethyl-2-[[(phenylacetyl)amino]methyl]-4-thiazolidinecarboxylic Acid, TRC

Product Code. 30386553
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386553 5 mg
30372428 1 mg
30385690 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386553 Supplier Toronto Research Chemicals Supplier No. TRCD4736855MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73184-06-2
Molecular Formula C15H20N2O3S
Molecular Weight (g/mol) 308.4
Synonym D-alpha-Benzylpenilloic Acid, (2R,4S)-5,5-Dimethyl-2-[[(2-phenylacetyl)amino]methyl]-4-thiazolidinecarboxylic Acid
SMILES CC1(C)S[C@H](CNC(=O)Cc2ccccc2)N[C@H]1C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H20N2O3S/c1-15(2)13(14(19)20)17-12(21-15)9-16-11(18)8-10-6-4-3-5-7-10/h3-7,12-13,17H,8-9H2,1-2H3,(H,16,18)(H,19,20)/t12-,13+/m1/s1
Formula Weight 308.1195
Chemical Name or Material (2R-trans)-5,5-Dimethyl-2-(((phenylacetyl)amino)methyl)-4-thiazolidinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.