missing translation for 'onlineSavingsMsg'
Learn More

(2R)-Tetrahydro-2-furanmethanamine Hydrochloride, TRC

Product Code. 30423789
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30423789 1 g
30412896 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30423789 Supplier Toronto Research Chemicals Supplier No. TRCT2935551G
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7175-80-6
Molecular Formula C5H11NO . HCl
Molecular Weight (g/mol) 137.61
Synonym (R)-Tetrahydro-2-furanmethanamine Hydrochloride, (R)-Tetrahydro-furfurylamine Hydrochloride, (-)-(R)-Tetrahydrofurfurylamine Hydrochloride, (2R)-2-Aminomethyltetrahydrofuran Hydrochloride, (2R)-Tetrahydrofuran-2-methanamine Hydrochloride, (R)-(-)-Tetrahydrofurfurylamine Hydrochloride, (R)-(Tetrahydrofuran-2-yl)methanamine Hydrochloride, (R)-1-(Tetrahydrofuran-2-yl)methylamine Hydrochloride, (R)-Tetrahydrofurfurylamine Hydrochloride, [((R)-Tetrahydrofuran-2-yl)methyl]amine Hydrochloride, [(R)-1-(Tetrahydro-furan-2-yl)]-methylamine Hydrochloride, [[(2R)-Tetrahydro-2-furanyl]methyl]amine Hydrochloride
SMILES NC[C@H]1CCCO1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H11NO.ClH/c6-4-5-2-1-3-7-5;/h5H,1-4,6H2;1H/t5-;/m1./s1
Chemical Name or Material (2R)-Tetrahydro-2-furanmethanamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.