missing translation for 'onlineSavingsMsg'
Learn More

(2R)-Octyl-α-hydroxyglutarate, MedChemExpress

Product Code. 16436607
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16436607 5 mg 5mg
16483878 10 mg 10mg
16462886 25 mg 25mg
16441029 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16436607 Supplier Medchem Express Supplier No. HY103641

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (2R)-Octyl-α-hydroxyglutarate ((2R)-Octyl-2-HG) is a modified form of D-isomer 2-Hydroxyglutarate.

(2R)-Octyl-α-hydroxyglutarate ((2R)-Octyl-2-HG) is a modified form of D-isomer 2-Hydroxyglutarate.

Specifications

Chemical Name or Material (2R)-Octyl-α-hydroxyglutarate
Physical Form Solid
CAS 1391194-67-4
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C13H24O5
For Use With (Application) Cancer-programmed cell death
Synonym (2R)-Octyl-2-HG
Solubility Information DMSO : 100 mg/mL (384.13 mM; Need ultrasonic)
SMILES O=C(OCCCCCCCC)[C@H](O)CCC(O)=O
Molecular Weight (g/mol) 260.33
Formula Weight 260.33
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.