missing translation for 'onlineSavingsMsg'
Learn More

(2R)-Lacosamide-d3 (O-methyl-d3), CDN

Product Code. 30326218
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Product Code. Quantity unitSize
30326218 0.01 g 0.01g
30324593 0.005 g 0.01g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30326218

Brand: CDN CDND72710.01G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lacosamide-d3
CAS 1217689-95-6
Molecular Formula C13 D3 H15 N2 O3
Quantity 0.01 g
InChI Formula InChI=1 S/C13H18N2O3/c1-10(16)15-12(9-18-2)13(17)14-8-11-6-4-3-5-7-11/h3-7,12 H,8-9H2,1-2H3,(H,14,17)(H,15,16)/t12-/m1/s1/i2D3
SMILES [2 H]C([2 H])([2 H])OC[C@@H](NC(=O)C)C(=O)NCc1ccccc1
IUPAC Name (2 R)-2-acetamido-N-benzyl-3-(trideuteriomethoxy)propanamide
Molecular Weight (g/mol) 253.31
Formula Weight 253.1506 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.