missing translation for 'onlineSavingsMsg'
Learn More

(2R-cis)-1-[2-(Hydroxymethyl)-1,3-oxathiolan-5-yl]-2,4(1H,3H)-pyrimidinedione, TRC

Product Code. 30137934
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137934 250 mg 250mg
30136335 100 mg 100mg
17994725 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137934 Supplier Toronto Research Chemicals Supplier No. TRCL172535250MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145986-07-8
Molecular Formula C8 H10 N2 O4 S
Molecular Weight (g/mol) 230.24
Synonym 2,4(1H,3H)-Pyrimidinedione, 1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, (2R-cis)- (9CI), 1,3-Oxathiolane, 2,4(1H,3H)-pyrimidinedione deriv., 1-[(2R,5S)-2-(Hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidine-2,4(1H,3H)-dione, Lamivudine Imp. J (EP)
IUPAC Name 1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidine-2,4-dione
SMILES OC[C@@H]1O[C@@H](CS1)N2C=CC(=O)NC2=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H10N2O4S/c11-3-7-14-6(4-15-7)10-2-1-5(12)9-8(10)13/h1-2,6-7,11H,3-4H2,(H,9,12,13)/t6-,7+/m0/s1
Formula Weight 230.0361
Chemical Name or Material (2R-cis)-1-[2-(Hydroxymethyl)-1,3-oxathiolan-5-yl]-2,4(1H,3H)-pyrimidinedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.