missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2R)-Arimoclomol Maleic Acid, TRC
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
Specifications
Specifications
| Chemical Name or Material | (2R)-Arimoclomol Maleic Acid |
| CAS | 289893-26-1 |
| Molecular Formula | C18H24ClN3O7 |
| Quantity | 0.5 mg |
| Synonym | N-{[(2R)-2-Hydroxy-3-piperidin-1-ylpropyl]oxy}pyridine-3-carboximidoyl Chloride 1-Oxide (2Z)-2-butenedioate, BRX 220 |
| InChI Formula | InChI=1S/C14H20ClN3O3.C4H4O4/c15-14(12-5-4-8-18(20)9-12)16-21-11-13(19)10-17-6-2-1-3-7-17;5-3(6)1-2-4(7)8/h4-5,8-9,13,19H,1-3,6-7,10-11H2;1-2H,(H,5,6)(H,7,8)/b;2-1-/t13-;/m1./s1 |
| SMILES | C1CCN(CC1)C[C@H](CO/N=C(/C2=C[N+](=CC=C2)[O-])\Cl)O.C(=C\C(=O)O)\C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;N-[(2R)-2-hydroxy-3-piperidin-1-ylpropoxy]-1-oxidopyridin-1-ium-3-carboximidoyl chloride |
| Molecular Weight (g/mol) | 429.85 |
| Formula Weight | 429.1303 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction