missing translation for 'onlineSavingsMsg'
Learn More

(2R)-Arimoclomol Maleic Acid, TRC

Product Code. 30335513
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30335513

Brand: Toronto Research Chemicals TRCA7712705MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-Arimoclomol Maleic Acid
CAS 289893-26-1
Molecular Formula C18H24ClN3O7
Quantity 5 mg
Synonym N-{[(2R)-2-Hydroxy-3-piperidin-1-ylpropyl]oxy}pyridine-3-carboximidoyl Chloride 1-Oxide (2Z)-2-butenedioate, BRX 220
InChI Formula InChI=1S/C14H20ClN3O3.C4H4O4/c15-14(12-5-4-8-18(20)9-12)16-21-11-13(19)10-17-6-2-1-3-7-17;5-3(6)1-2-4(7)8/h4-5,8-9,13,19H,1-3,6-7,10-11H2;1-2H,(H,5,6)(H,7,8)/b;2-1-/t13-;/m1./s1
SMILES C1CCN(CC1)C[C@H](CO/N=C(/C2=C[N+](=CC=C2)[O-])\Cl)O.C(=C\C(=O)O)\C(=O)O
IUPAC Name (Z)-but-2-enedioic acid;N-[(2R)-2-hydroxy-3-piperidin-1-ylpropoxy]-1-oxidopyridin-1-ium-3-carboximidoyl chloride
Molecular Weight (g/mol) 429.85
Formula Weight 429.1303
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.