missing translation for 'onlineSavingsMsg'
Learn More
Learn More
((2R,5S,10aS,10bS)-10b-Hydroxy-5-isopropyl-2-methyl-3,6-dioxooctahydro-2H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl)carbamic Acid Benzyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | ((2R,5S,10aS,10bS)-10b-Hydroxy-5-isopropyl-2-methyl-3,6-dioxooctahydro-2H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl)carbamic Acid Benzyl Ester |
| Analyte or Component Names | ((2R,5S,10aS,10bS)-10b-Hydroxy-5-isopropyl-2-methyl-3,6-dioxooctahydro-2H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl)carbamic Acid Benzyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H37 N O4 |
| InChI Formula | InChI=1S/C26H37NO4/c1-22(2,3)23(4,28)17-14-24-9-10-26(17,30-6)21-25(24)11-12-27-18(24)13-15-7-8-16(29-5)20(31-21)19(15)25/h7-8,17-18,21,27-28H,9-14H2,1-6H3/t17-,18-,21+,23-,24?,25+,26+/m1/s1 |
| Solution Type | Neat |
| SMILES | COc1ccc2C[C@H]3NCC[C@@]45[C@H](Oc1c24)[C@@]6(CCC35C[C@@H]6[C@@](C)(O)C(C)(C)C)OC |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 427.576 |
| Quantity | 5 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?