missing translation for 'onlineSavingsMsg'
Learn More

(2R,5R)-2,5-Hexanediol, 95%, 98% ee, Thermo Scientific™

Product Code. 10752833 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10752833 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10752833 Supplier Thermo Scientific Acros Supplier No. 363200010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 17299-07-9
CAS Min % 94.0
CAS Max % 100.0
Melting Point 50°C to 53°C
Color White to Yellow
Boiling Point 212°C to 215°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C6H14O2
Linear Formula CH3CH(OH)CH2CH2CH(OH)CH3
Quantity 1 g
Specific Rotation -34.5
Synonym 2r,5r-2,5-hexanediol, 2r,5r-hexane-2,5-diol, 2,5-hexanediol, 2r,5r, 2r,5r---2,5-hexanediol, hx2, 2r,5r-hexanediol, 2r,5r-2,5-dihydroxyhexane, 2r,5r-2,5-hexanediol sum of enantiomers, gc
InChI Key OHMBHFSEKCCCBW-PHDIDXHHSA-N
SMILES CC(CCC(C)O)O
IUPAC Name (2R,5R)-hexane-2,5-diol
Molecular Weight (g/mol) 118.18
PubChem CID 2733360
Formula Weight 118.18
Percent Purity 95%,98% ee
Physical Form Crystalline Powder or Needles
Chemical Name or Material (2R, 5R)-2, 5-Hexanediol, 98% ee
Show More Show Less
compliance-icons
Product Identifier
  • (2R,5R)-2,5-Hexanediol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.