missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2R,4R)-APDC, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
A highly selective and relatively potent group II metabotropic glutamate receptor agonist. EC50 values are 0.4, 0.4, > 100, > 100, > 300 and > 300 μM for human mGlu2, mGlu3, mGlu1, mGlu5, mGlu4 and mGlu7 receptors respectively. Centrally active following systemic administration in vivo.
Specifications
Quantity | 10 mg |
Synonym | 2r,4r-apdc, 2r,4r-4-aminopyrrolidine-2,4-dicarboxylic acid, chembl40086, 2r,4r-4-aminopyrrolidine-2,4-dicarboxylate, 2r, 4r-apdc monohydrate, 4-amino-2,4-pyrrolidinedicarboxylic acid, d06tod, d0ek6p, 2r, 4r-apdc |
Formula Weight | 174.16 |
Percent Purity | >98% |
Chemical Name or Material | (2R, 4R)-APDC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'