missing translation for 'onlineSavingsMsg'
Learn More

(2R,4R)-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol 4-(4-Methylbenzenesulfonate), TRC

Product Code. 30386511
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386511 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386511 Supplier Toronto Research Chemicals Supplier No. TRCD43702510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R,4R)-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol 4-(4-Methylbenzenesulfonate)
CAS 1610851-15-4
Molecular Formula C21 H20 Cl2 N2 O5 S
Quantity 10 mg
Synonym (2R,4R)-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolane-4-methanol 4-(4-Methylbenzenesulfonate)
InChI Formula InChI=1S/C21H20Cl2N2O5S/c1-15-2-5-18(6-3-15)31(26,27)29-12-17-11-28-21(30-17,13-25-9-8-24-14-25)19-7-4-16(22)10-20(19)23/h2-10,14,17H,11-13H2,1H3/t17-,21+/m1/s1
SMILES Cc1ccc(cc1)S(=O)(=O)OC[C@H]2CO[C@@](Cn3ccnc3)(O2)c4ccc(Cl)cc4Cl
IUPAC Name [(2R,4R)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 483.365
Formula Weight 482.047
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.