missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2R,3S)-N-Benzoyl-3-phenyl Isoserine, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | (2R,3S)-N-Benzoyl-3-phenyl Isoserine |
| CAS | 132201-33-3 |
| Molecular Formula | C16 H15 N O4 |
| Quantity | 100 mg |
| Synonym | Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, (αR,βS)-, Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, [R-(R*,S*)]-, (2R,3S)-3-(Benzoylamino)-2-hydroxy-3-phenylpropionic acid, (2R,3S)-N-Benzoyl-3-phenylisoserine, N-Benzoyl-(2R,3S)-3-phenylisoserine |
| InChI Formula | InChI=1S/C16H15NO4/c18-14(16(20)21)13(11-7-3-1-4-8-11)17-15(19)12-9-5-2-6-10-12/h1-10,13-14,18H,(H,17,19)(H,20,21)/t13-,14+/m0/s1 |
| SMILES | O[C@H]([C@@H](NC(=O)c1ccccc1)c2ccccc2)C(=O)O |
| IUPAC Name | (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoic acid |
| Molecular Weight (g/mol) | 285.29 |
| Formula Weight | 285.1001 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction