missing translation for 'onlineSavingsMsg'
Learn More

(2R,3R)-(-)-2,3-Butanediol, 98+%, 99+% ee

Product Code. 10509823 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10509823 5 g Glass bottle 5g
10523581 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10509823 Supplier Thermo Scientific Acros Supplier No. 217980050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24347-58-8 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24347-58-8
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00064267
InChI Key OWBTYPJTUOEWEK-UHFFFAOYNA-N
Synonym 2r,3r-butane-2,3-diol, r,r-2,3-butanediol, 2r,3r---2,3-butanediol, r,r-butane-2,3-diol, 2r,3r-2,3-butanediol, r,r-2,3-butylene glycol, r,r---butane-2,3-diol, r,r---2,3-butylene glycol, unii-or02b2286a, r,r---2,3-butanediol
PubChem CID 225936
ChEBI CHEBI:16982
IUPAC Name (2R,3R)-butane-2,3-diol
SMILES CC(O)C(C)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 16.0°C
Color Colorless
Density 0.9800g/mL
Boiling Point 77.3°C to 77.4°C (10.0 mmHg)
Flash Point 85°C
Enantiomeric Excess 99.0% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(OH)CH(OH)CH3
Refractive Index 1.4309 to 1.4329
Quantity 5 g
Beilstein 01, I, 250
Fieser 16,52
Merck Index 15, 1571
Specific Gravity 0.98
Specific Rotation Condition − 13.00 (20.00°C neat)
Specific Rotation − 13.00
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and diisopropyl ether
Formula Weight 90.12
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material (2R, 3R)-(-)-2, 3-Butanediol, ≥99% ee
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.