missing translation for 'onlineSavingsMsg'
Learn More

(2R)-2-Oxiranylmethyl Ester Butanoic Acid, TRC

Product Code. 30413934
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413934 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413934 Supplier Toronto Research Chemicals Supplier No. TRCO8470905G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-2-Oxiranylmethyl Ester Butanoic Acid
CAS 60456-26-0
Molecular Formula C7H12O3
Quantity 5 g
Synonym (R)-Oxiranylmethyl Ester Butanoic Acid, (-)-Glycidyl Butyrate, (2R)-(-)-Glycidyl Butyrate, (R)-(-)-Glycidyl Butyrate, (R)-Glycidyl Butyrate, (R)-Glycidyl Ester Butyric Acid
InChI Formula InChI=1S/C7H12O3/c1-2-3-7(8)10-5-6-4-9-6/h6H,2-5H2,1H3/t6-/m1/s1
SMILES CCCC(=O)OC[C@H]1CO1
IUPAC Name [(2R)-oxiran-2-yl]methyl butanoate
Molecular Weight (g/mol) 144.17
Formula Weight 144.0786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.