missing translation for 'onlineSavingsMsg'
Learn More

(2R)-2-Hydroxyglutaric Acid Disodium Salt, TRC

Product Code. 30136392
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136392 25 mg 25mg
30135966 100 mg 100mg
30012727 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136392 Supplier Toronto Research Chemicals Supplier No. TRCH94256525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103404-90-6
Molecular Formula C5 H6 O5 . 2 Na
Molecular Weight (g/mol) 192.08
Synonym Pentanedioic acid, 2-hydroxy-, sodium salt (1:2), (2R)-, Pentanedioic acid, 2-hydroxy-, disodium salt, (2R)- (9CI), Pentanedioic acid, 2-hydroxy-, disodium salt, (R)-, (2R)-2-Hydroxypentanedioic acid disodium salt, (R)-(-)-2-Hydroxyglutaric acid disodium salt, (R)-2-Hydroxyglutaric acid disodium salt, (R)-2-Hydroxypentanedioic acid disodium salt, (R)-α-Hydroxyglutaric acid disodium salt, D-α-Hydroxyglutaric acid disodium salt
IUPAC Name disodium;(2R)-2-hydroxypentanedioate
SMILES [Na+].[Na+].O[C@H](CCC(=O)[O-])C(=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H8O5.2Na/c6-3(5(9)10)1-2-4(7)8;;/h3,6H,1-2H2,(H,7,8)(H,9,10);;/q;2*+1/p-2/t3-;;/m1../s1
Formula Weight 192.0011
Chemical Name or Material (2R)-2-Hydroxyglutaric Acid Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.