missing translation for 'onlineSavingsMsg'
Learn More

(2R)-2-Hydroxyglutaric Acid Disodium Salt-13C5, TRC

Product Code. 30390600
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390600 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390600 Supplier Toronto Research Chemicals Supplier No. TRCH9425680.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-2-Hydroxyglutaric Acid Disodium Salt-13C5
CAS 1648909-80-1
Molecular Formula 13C5H6Na2O5
Quantity 0.5 mg
Synonym (2R)-2-Hydroxypentanedioic Acid Disodium Salt-13C5, (R)-2-Hydroxyglutaric Acid Disodium Salt-13C5, (R)-α-Hydroxyglutaric Acid Disodium Salt-13C5, D-2-Hydroxyglutaric Acid Disodium Salt-13C5
InChI Formula InChI=1S/C5H8O5.2Na/c6-3(5(9)10)1-2-4(7)8;;/h3,6H,1-2H2,(H,7,8)(H,9,10);;/q;2*+1/p-2/t3-;;/m1../s1/i1+1,2+1,3+1,4+1,5+1;;
SMILES O=[13C]([13CH2][13CH2][13C@](O)([H])[13C](O[Na])=O)O[Na]
IUPAC Name sodium (R)-2-hydroxypentanedioate-1,2,3,4,5-13C5
Molecular Weight (g/mol) 197.04
Formula Weight 197.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.