Learn More
Greiner Bio-One 2mL Polypropylene Microcentrifuge Tubes
Description
- Material: Polypropylene
- For single use only
- With attached cap and graduation
- Available in different colors
- Free of detectable DNase/RNase, human DNA and pyrogens
- Suitable for Eppendorf centrifuges and system
Service Plan
No
Warranty
No
Specifications
Specifications
| Color | Natural |
| Material | Polypropylene |
| Volume (Metric) | 2mL |
| Sterility | Non-sterile |
| Graduated | Yes |