missing translation for 'onlineSavingsMsg'
Learn More

Sarstedt™ 2mL Polypropylene Micro Tube

Product Code. 12705493
Encompass
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
12705493 Sterile Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12705493 Supplier Sarstedt™ Supplier No. 72.694.006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Screw cap micro tubes without printing are ideal for applications in which an accurate inspection of the sample is particularly important. The transparent polyethylene facilitates the visual inspection of the content.

  • Ideal centrifuge tubes as well as transport or storage vessels, especially for buffers loading solutions, media or other higher volume reagents
  • Precise thread and a flexible O-ring ensure the special sealing security of the screw cap
  • Autoclavable at 121°C
  • Stable skirted base provides secure footing on the laboratory bench, even without a rack
  • Special knurling - one-handed screwing is easily possible by locking into the special rack
  • Graduated in white

Specifications

Cap Style Screw cap
Closure Color Transparent
Closure Material Polypropylene
Autoclavable Autoclavable (121°C)
Capacity (Metric) 2 mL
Color Transparent
Material Polypropylene
Packaging 1000/box
Sterility Sterile
Graduated Yes
Diameter (Metric) 10.8 mm
Length (Metric) 44 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.