Learn More
Thermo Scientific™ 2mL Amber Vials
Product Code. 15348036
Change view
Click to view available options
Color:
Amber
Unit Size:
Pack of 100
Product Code. Color unitSize
15348036 Amber Pack of 100
1 options
Product Code. 15348036 Supplier Thermo Scientific™ Supplier No. C500076W
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use certified clean, low particle, low background vials for your high sensitivity LC/MS and GC/MS applications.
  • 100 vials and 100 caps with pre-assembled septa
  • Reusable two-compartment trays protect vials and closures while keeping matching supplies together
  • Clear trays make it easy to keep track of available supplies without opening containers
  • 2mL
TRUSTED_SUSTAINABILITY
Specifications
Color Amber
Septum SLIT T
No. per Pack 1000