missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ RMR-Tre

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
RMR-Tre is a far-red fluorogenic trehalose specific probe for live mycobacteria imaging. It enables fast, no-wash, low-background fluorescence detection of live mycobacterial outer membrane. RMR-Tre contains a molecular rotor fluorophore that brightly emits signal in the far-red region upon trehalose-guided metabolic integration into the mycomembrane. It exhibited up to a 100- fold enhancement in Mycobacterium tuberculosis labeling compared to existing fluorogenic trehalose probes. Excitation and emission maxima (λ) are 549 nm and 571 nm, respectively.
Specifications
Specifications
| Chemical Name or Material | 3-(6-(5-(2,2-Dicyanovinyl)thiophen-2-yl)-3,4-dihydroquinolin-1(2H)-yl)-N-(((2R,3S,4S,5R,6R)-3,4,5-trihydroxy-6-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)methyl)propanamide |
| Quantity | 2 mg |
| Target | Fluorescent Probes for Imaging Bacteria |
| Molecular Formula | C32H38N4O11S |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction