missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NDT 30805
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
NDT 30805 is a potent NLRP3 inflammasome inhibitor. Inhibits IL-1β release in peripheral blood mononuclear cell assay, ASC speck assay and in whole blood (IC50 values are 13 nM, 34 nM and 1500 nM). Inhibits Nigericin (Cat. No. 4312) and ATP-induced IL-1β release. Soluble and uncharged at physiological pH.
Specifications
Specifications
| Chemical Name or Material | 2-((1,2,3,5,6,7-Hexahydro-s-indacen-4-yl)amino)-5-(pyridin-2-ylmethyl)-[1,2,4]triazolo[1,5-a]pyrimidine-7(4H)-thione |
| Quantity | 2 mg |
| Target | Inflammasomes Inhibitors |
| Molecular Formula | C23H22N6S |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction