missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ hCAII Degrader 11
Click to view available options
:
2 mg
Description
hCAII Degrader 11 is a potent human carbonic anhydrase II (hCAII) Degrader (PROTAC™; Dmax = 100% at 5 nM, DC50 = 0.5 nM). PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license. Sold with kind permission of Professor Cohen and Professor Burkart, Department of Chemistry and Biochemistry, University of California, USA.
Specifications
Specifications
| Chemical Name or Material | 4-(Aminosulfonyl)-N-[12-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]amino]dodecyl]benzamide |
| CAS | 2892306-56-6 |
| Quantity | 2 mg |
| Target | Active Degraders |
| Molecular Formula | C32H41N5O7S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction