missing translation for 'onlineSavingsMsg'
Learn More

2H-Pyran-2-one, 97%

Product Code. 10236802 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10236802 5 g Glass bottle 5g
10464052 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10236802 Supplier Thermo Scientific Acros Supplier No. 376810050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 504-31-4 | C5H4O2 | 96.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-31-4
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.09
MDL Number MFCD00006639
InChI Key ZPSJGADGUYYRKE-UHFFFAOYSA-N
Synonym 2h-pyran-2-one, alpha-pyrone, 2-pyranone, 2-pyrone, coumalin, pyrone, .alpha.-pyrone, unii-8ww45i202v, 2-oxo-2h-pyran, oxidopyrylium
PubChem CID 68154
ChEBI CHEBI:37965
IUPAC Name pyran-2-one
SMILES C1=CC(=O)OC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 8°C to 9°C
Color Yellow to Brown
Density 1.2100g/mL
Boiling Point 104°C (30.0 mmHg)
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5280 to 1.531
Quantity 5 g
Specific Gravity 1.21
Formula Weight 96.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2H-Pyran-2-one
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.