missing translation for 'onlineSavingsMsg'
Learn More

2H,3H-Perfluoropentane, tech. 90%, Thermo Scientific Chemicals

Product Code. p-7025905
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

Product Code. 11420187

Brand: Thermo Scientific Alfa Aesar B21633.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

Used as a zero ozone-depletion alternative to hydro chlorofluorocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a zero ozone-depletion alternative to hydro chlorofluorocarbons.

Solubility
Soluble in water (140 mg/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138495-42-8
Molecular Formula C5H2F10
Molecular Weight (g/mol) 252.06
MDL Number MFCD00798129
InChI Key RIQRGMUSBYGDBL-UHFFFAOYNA-N
Synonym 2h,3h-decafluoropentane, vertrel xf, 2h,3h-perfluoropentane, hfc-4310mee, 1,1,1,2,3,4,4,5,5,5-decafluoropentane, 2,3-dihydroperfluoropentane, pentane, 1,1,1,2,2,3,4,5,5,5-decafluoro, hfc-43-10mee, 2,3-dihydrodecafluoropentane, hfc-43
PubChem CID 86240
IUPAC Name 1,1,1,2,2,3,4,5,5,5-decafluoropentane
SMILES FC(C(F)C(F)(F)C(F)(F)F)C(F)(F)F

Specifications

Melting Point -80°C
Density 1.6
Boiling Point 54°C
Flash Point None
Linear Formula CF3CHFCHFCF2CF3
Refractive Index <1.3000
Quantity 5 g
Solubility Information Soluble in water (140mg/L at 25°C).
Formula Weight 252.05
Percent Purity 90%
Grade Technical
Chemical Name or Material 2H,3H-Perfluoropentane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2H,3H-Perfluoropentane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 2H,3H-Perfluoropentane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.