missing translation for 'onlineSavingsMsg'
Learn More

2H-1,2,3-Triazole-4-carboxylic Acid, TRC

Product Code. 30395732
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395732 1 g
30401519 250 mg
30398427 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395732 Supplier Toronto Research Chemicals Supplier No. TRCH2090361G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 573714-78-0
Molecular Formula C3 H3 N3 O2
Molecular Weight (g/mol) 113.075
IUPAC Name 1H-triazole-4-carboxylic acid
SMILES OC(=O)c1c[nH]nn1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C3H3N3O2/c7-3(8)2-1-4-6-5-2/h1H,(H,7,8)(H,4,5,6)
Formula Weight 113.023
Chemical Name or Material 2H-1,2,3-Triazole-4-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.