missing translation for 'onlineSavingsMsg'
Learn More

(2E)-Demethoxy Curcumin, TRC

Product Code. 30012248
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012248 25 mg 25mg
30006740 10 mg 10mg
30007935 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012248 Supplier Toronto Research Chemicals Supplier No. TRCD23081525MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22608-11-3
Molecular Formula C20 H18 O5
Molecular Weight (g/mol) 338.35
Synonym Demethoxycurcumin, E,E-Desmethoxycurcumin, 1,6-Heptadiene-3,5-dione, 1-(4-hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-, (1E,6E)-, 1,6-Heptadiene-3,5-dione, 1-(4-hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-, (E,E)-, 1,6-Heptadiene-3,5-dione, 1-(4-hydroxy-3-methoxyphenyl)-7-(p-hydroxyphenyl)- (8CI), (1E,6E)-1-(4-Hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, (1E,6E)-1-(4-Hydroxyphenyl)-7-(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Curcumin II, Demethoxycurcumin, Desmethoxycurcumin, Monodemethoxycurcumin, 1,6-Heptadiene-3,5-dione, 1-(4-hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-, (1E,6E)-, 1,6-Heptadiene-3,5-dione, 1-(4-hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-, (E,E)-, 1,6-Heptadiene-3,5-dione, 1-(4-hydroxy-3-methoxyphenyl)-7-(p-hydroxyphenyl)- (8CI), (1E,6E)-1-(4-Hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, (1E,6E)-1-(4-Hydroxyphenyl)-7-(4-hydroxy-3-methoxyphenyl)-1,6-heptadiene-3,5-dione, Curcumin II, Demethoxycurcumin, Desmethoxycurcumin, Monodemethoxycurcumin
IUPAC Name (1E,6E)-1-(4-hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione
SMILES COc1cc(\C=C\C(=O)CC(=O)\C=C\c2ccc(O)cc2)ccc1O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H18O5/c1-25-20-12-15(6-11-19(20)24)5-10-18(23)13-17(22)9-4-14-2-7-16(21)8-3-14/h2-12,21,24H,13H2,1H3/b9-4+,10-5+
Formula Weight 338.1154
Percent Purity >95%
Chemical Name or Material Demethoxycurcumin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.