missing translation for 'onlineSavingsMsg'
Learn More

(2E,5E)-2,5-Bis[(4-(diethylamino)phenyl)methylene]cyclopentanone, 95%, Thermo Scientific™

Product Code. 10197682 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10197682 5 g 5g
10187682 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10197682 Supplier Thermo Scientific Acros Supplier No. 375010050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261360-66-1
Molecular Formula C27H34N2O
Molecular Weight (g/mol) 402.58
MDL Number MFCD06658953
InChI Key KKLBPVXKMBLCQX-BLVCXSLXSA-N
Synonym cyclopentanone, 2,5-bis 4-diethylamino phenyl methylene, 2e,5e-2,5-bis 4-diethylamino phenyl methylene cyclopentanone, 2,5-bis 4-diethylamino benzylidene cyclopentanone, 2,5-bis 4-diethylaminobenzylidene cyclopentanone 17, 2,5-bis e-4-diethylamino benzylidene cyclopentanone, 2e 5e-2,5-bis 4-diethylamino benzylidene cyclopentanone, 2e,5e-2,5-bis 4-diethylamino benzylidene cyclopentanone, 2,5-di e-1-4-diethylaminophenyl methylidene-1-cyclopentanone, 2e,5e-2,5-bis 4-diethylamino phenyl methylidene cyclopentan-1-one
PubChem CID 5702661
IUPAC Name (2E,5E)-2,5-bis[[4-(diethylamino)phenyl]methylidene]cyclopentan-1-one
SMILES CCN(CC)C1=CC=C(C=C1)C=C2CCC(=CC3=CC=C(C=C3)N(CC)CC)C2=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 186°C to 189°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 402.58
Percent Purity 95%
Physical Form Powder
Chemical Name or Material (2E, 5E)-2, 5-Bis[(4-(diethylamino)phenyl)methylene]cyclopentanone
Show More Show Less
compliance-icons
Product Identifier
  • (2E,5E)-2,5-Bis[(4-(diethylamino)phenyl)-methylene]cyclopentanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.