missing translation for 'onlineSavingsMsg'
Learn More

(2E,4E)-5-[(2S,3S)-3-[(3S)-3,4-Dihydroxy-4-methylpentyl]-3-methyl-2-oxiranyl]-3-methyl-2,4-pentadienoic Acid Methyl Ester, TRC

Product Code. 30385231
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385231 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385231 Supplier Toronto Research Chemicals Supplier No. TRCD45377525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2E,4E)-5-[(2S,3S)-3-[(3S)-3,4-Dihydroxy-4-methylpentyl]-3-methyl-2-oxiranyl]-3-methyl-2,4-pentadienoic Acid Methyl Ester
CAS 153651-94-6
Molecular Formula C14 H19 N3 O5 S
Quantity 25 mg
Synonym [2S-[2alpha(2E,4E),3beta(R*)]]-5-[3-(3,4-Dihydroxy-4-methylpentyl)-3-methyloxiranyl]-3-methyl-2,4-Pentadienoic acid
InChI Formula InChI=1S/C14H19N3O5S/c1-6(18)9-12(20)17-10(14(21)22)8(23-13(9)17)5-16-4-2-3-7(16)11(15)19/h6-7,9,13,18H,2-5H2,1H3,(H2,15,19)(H,21,22)/t6-,7+,9+,13-/m1/s1
SMILES C[C@@H](O)[C@@H]1[C@H]2SC(=C(N2C1=O)C(=O)O)CN3CCC[C@H]3C(=O)N
IUPAC Name (5R,6S)-3-[[(2S)-2-carbamoylpyrrolidin-1-yl]methyl]-6-[(1R)-1-hydroxyethyl]-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid
Molecular Weight (g/mol) 341.383
Formula Weight 341.105
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.