missing translation for 'onlineSavingsMsg'
Learn More

(2E)-3-Methyl-1,3-benzothiazol-2(3H)-one Hydrazone Hydrochloride (>85%), TRC

Product Code. 30138592
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138592 1 g 1g
30010515 100 mg 100mg
30003497 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138592 Supplier Toronto Research Chemicals Supplier No. TRCE9752481G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38894-11-0
Molecular Formula C8 H9 N3 S . H Cl . H2 O
Molecular Weight (g/mol) 233.718
Synonym Sawicki Reagent;
IUPAC Name (3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrate;hydrochloride
SMILES O.Cl.CN1C(=NN)Sc2ccccc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H9N3S.ClH.H2O/c1-11-6-4-2-3-5-7(6)12-8(11)10-9;;/h2-5H,9H2,1H3;1H;1H2
Formula Weight 233.039
Chemical Name or Material (2E)-3-Methyl-1,3-benzothiazol-2(3H)-one Hydrazone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.