missing translation for 'onlineSavingsMsg'
Learn More

(2E)-3-(4-Hydroxy-3,5-dimethoxyphenyl)-2-propenoic Acid, TRC

Product Code. 30389864
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389864 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389864 Supplier Toronto Research Chemicals Supplier No. TRCH80528050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-Sinapic Acid
CAS 7362-37-0
Molecular Formula C11 H12 O5
Quantity 50 g
Synonym 2-Propenoic acid, 3-(4-hydroxy-3,5-dimethoxyphenyl)-, (2E)-, 2-Propenoic acid, 3-(4-hydroxy-3,5-dimethoxyphenyl)-, (E)-, Cinnamic acid, 4-hydroxy-3,5-dimethoxy-, (E)- (8CI), (2E)-3-(4-Hydroxy-3,5-dimethoxyphenyl)-2-propenoic acid, (E)-3,5-Dimethoxy-4-hydroxycinnamic acid, (E)-Sinapic acid, 3,5-Dimethoxy-4-hydroxy-trans-cinnamic acid, E-Sinapinic acid, trans-4-Hydroxy-3,5-dimethoxycinnamic acid, trans-Sinapic acid, trans-Sinapinic acid
InChI Formula InChI=1S/C11H12O5/c1-15-8-5-7(3-4-10(12)13)6-9(16-2)11(8)14/h3-6,14H,1-2H3,(H,12,13)/b4-3+
SMILES COc1cc(\C=C\C(=O)O)cc(OC)c1O
IUPAC Name (E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 224.21
Formula Weight 224.0685
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.