missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2E)-1-[2-[(2RS)-2-Hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylprop-2-en-1-one Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(2E)-1-[2-[(2RS)-2-Hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylprop-2-en-1-one Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (2E)-1-[2-[(2RS)-2-Hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylprop-2-en-1-one Hydrochloride |
| CAS | 2468200-18-0 |
| Analyte or Component Names | (2E)-1-[2-[(2RS)-2-Hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylprop-2-en-1-one Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H25 N O3 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Propafenone Hydrochloride Imp. B (EP), Propafenone Imp. B (EP), Propafenone USP Related Compound B, Propafenone USP RC B, (2E)-1-[2-[(2RS)-2-Hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylprop-2-en-1-one Hydrochloride, Propafenone Hydrochloride Imp. B (EP) as Hydrochloride, Propafenone Related Compound B as Hydrochloride, Propafenone Hydrochloride Impurity B as Hydrochloride, Propafenone Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C21H25NO3.ClH/c1-2-14-22-15-18(23)16-25-21-11-7-6-10-19(21)20(24)13-12-17-8-4-3-5-9-17;/h3-13,18,22-23H,2,14-16H2,1H3;1H/b13-12+; |
| SMILES | Cl.CCCNCC(O)COc1ccccc1C(=O)\C=C\c2ccccc2 |
| IUPAC Name | (E)-1-[2-[2-hydroxy-3-(propylamino)propoxy]phenyl]-3-phenylprop-2-en-1-one;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction