Learn More
Thermo Scientific™ Thermo Scientific™ 2D Tank Liner, ASI™ 26 film
Single-use tank liners reduce cleaning steps common to re-usable tanks.
Brand: Thermo Scientific™ B100048-I
Description
The Thermo Scientific™ 2D Tank Liner System without bottom drain is composed of Thermo Scientific™ ASI™ 26 Polyethylene Film, a durable 5.0 mL film, with dimensions optimized for Thermo Scientific™ drums and commonly-used, industry standard cylindrical tanks. Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
- Minimizes bioburden due to construction under sterile cGMP conditions
- Eliminates post-use cleaning steps required with reusable containers, helping to reduce cross-contamination risks and reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
| 50 L | |
| Store at room temperature. | |
| 40 per case | |
| Irradiated | |
| HyPerforma™ | |
| ASI™ 26 |
| Polyethylene | |
| 33 cm x 68.6 cm (13 in. x 27 in.) | |
| SV30111 | |
| Room Temperature | |
| 0 ports | |
| No Tubing |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.