missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors, injection site

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.

Brand:  Thermo Scientific™ SS00027-I

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 17875726

  • £568.94 / Pack of 30
Estimated Shipment: 15-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.

Features
• Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
• Constructed in an ISO 7 clean room under cGMP conditions

Applications
• Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
• Preparing and storing buffer, culture media, and process liquids
• Collecting chromatography feeds and fractions
• Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters (used for bioburden reduction )—sterilization grade filters, vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style ports, needle-free ports
• Clamps—sliders, pinch clamps, crimp bands

BPC port specifications:
• Line 1: Luer lock insert connection with 4.8 mm (3/16 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
• Line 2: Luer lock body connection with 4.8 mm (3/16 in.) barb, cap, and slide clamp. Tubing: EVA + TPE 374
• Line 3: Tubing: EVA, injection port

Specifications

Specifications

1 L
Store at room temperature.
One 4-mil. poly bag
Irradiated
Labtainer™
ASI 26/77
Polyethylene
32 x 14.7 cm (12.6 x 5.8 in.)
50/Cs.
Room Temperature
Luer Lock Body Connector and Injection Site
TPE and EVA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers