missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Reusable white polycarbonate box holds 100 cryogenic vials in a 10 x 10 array
- Only for use with 1 - 2 mL internally threaded cryogenic vials
- Ridges on sides of box make them easier to grip while wearing gloves
Specifications
Specifications
| Type | Cryo Storage Box |
| Holds | 100 Vials |
| Tube Array | 10 x 10 |
| Weight (Metric) | 12 kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?