missing translation for 'onlineSavingsMsg'
Learn More

(2bS,5S,8S,9S)-5-Hydroxy-9-methyl-6-methylene-11-oxododecahydro-1aH-2,9-(epoxymethanooxy)-5,7a-methanocyclohepta[1,2]indeno[5,6-b]oxirene-8-carboxylic Acid, TRC

Product Code. 30398628
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398628 2.5 mg 2.5mg
30401494 25 mg 25mg
30391659 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398628 Supplier Toronto Research Chemicals Supplier No. TRCH5160952.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2bS,5S,8S,9S)-5-Hydroxy-9-methyl-6-methylene-11-oxododecahydro-1aH-2,9-(epoxymethanooxy)-5,7a-methanocyclohepta[1,2]indeno[5,6-b]oxirene-8-carboxylic Acid
Analyte or Component Names (2bS,5S,8S,9S)-5-Hydroxy-9-methyl-6-methylene-11-oxododecahydro-1aH-2,9-(epoxymethanooxy)-5,7a-methanocyclohepta[1,2]indeno[5,6-b]oxirene-8-carboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C19 H22 O7
InChI Formula InChI=1S/C19H22O7/c1-7-5-18-6-19(7,23)4-3-8(18)9-10(11(18)15(20)21)17(2)14-13(24-14)12(9)25-16(22)26-17/h8-14,23H,1,3-6H2,2H3,(H,20,21)/t8-,9?,10?,11+,12?,13?,14?,17-,18?,19?/m0/s1
Solution Type Neat
SMILES C[C@@]12OC(=O)OC(C3OC13)C4[C@@H]5CCC6(O)CC5(CC6=C)[C@H](C24)C(=O)O
IUPAC Name (3S,10S,12S)-6-hydroxy-12-methyl-7-methylidene-17-oxo-14,16,18-trioxahexacyclo[10.3.3.16,9.02,11.03,9.013,15]nonadecane-10-carboxylic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 362.374
Quantity 2.5 mg
Formula Weight 362.137
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.