missing translation for 'onlineSavingsMsg'
Learn More

2Beta-Hydroxy Medroxy Progesterone 17-Acetate (>90%), TRC

Product Code. 30401462
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401462 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401462 Supplier Toronto Research Chemicals Supplier No. TRCH94451525MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1072923-64-8
Molecular Formula C24H34O5
Quantity 25 mg
Synonym (2beta,6alpha)-17-(Acetyloxy)-2-hydroxy-6-methylpregn-4-ene-3,20-dione
InChI Formula InChI=1S/C24H34O5/c1-13-10-16-17(22(4)12-21(28)20(27)11-19(13)22)6-8-23(5)18(16)7-9-24(23,14(2)25)29-15(3)26/h11,13,16-18,21,28H,6-10,12H2,1-5H3/t13-,16+,17-,18-,21-,22+,23-,24-/m0/s1
SMILES C[C@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2CC[C@]3(OC(=O)C)C(=O)C)[C@@]4(C)C[C@H](O)C(=O)C=C14
IUPAC Name [(2S,6S,8R,9S,10R,13S,14S,17R)-17-acetyl-2-hydroxy-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 402.53
Formula Weight 402.2406
Chemical Name or Material 2beta-Hydroxy Medroxy Progesterone 17-Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.