missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2B4/CD244/SLAMF4 Antibody (MM0070-7D24), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
2B4/CD244/SLAMF4 Monoclonal antibody specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | 2B4/CD244/SLAMF4 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | MM0070-7D24 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL |
| Host Species | Mouse |
| Immunogen | Human recombinant 2B4/CD244/SLAMF4 (extracellular domain) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?