missing translation for 'onlineSavingsMsg'
Learn More

2B4/CD244/SLAMF4 Antibody (025), DyLight 488, Novus Biologicals™

Product Code. 18384741 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384741 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384741 Supplier Novus Biologicals Supplier No. NBP289232G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 2B4/CD244/SLAMF4
Applications Western Blot
Classification Monoclonal
Clone 25
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human 2B4/CD244/SLAMF4 (NP_057466.1; Met1-Arg221).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Myeloid derived Suppressor Cell, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 51744
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.