missing translation for 'onlineSavingsMsg'
Learn More

2b,3b-Epoxy-17-methyl-5a-androstan-17b-ol, TRC

Product Code. 30388013
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388013 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388013 Supplier Toronto Research Chemicals Supplier No. TRCE492755500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2b,3b-Epoxy-17-methyl-5a-androstan-17b-ol
CAS 968-55-8
Molecular Formula C20 H32 O2
Quantity 500 mg
Synonym (1S,3aS,3bR,5aS,6aR,7aS,8aS,8bS,10aS)-1,8a,10a-Trimethylhexadecahydro-1H-cyclopenta[7,8]phenanthro[2,3-b]oxiren-1-ol
InChI Formula InChI=1S/C20H32O2/c1-18-11-17-16(22-17)10-12(18)4-5-13-14(18)6-8-19(2)15(13)7-9-20(19,3)21/h12-17,21H,4-11H2,1-3H3/t12-,13+,14-,15-,16+,17-,18-,19-,20-/m0/s1
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4C[C@H]5O[C@H]5C[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (1S,2S,4S,6R,8S,11R,12S,15S,16S)-2,15,16-trimethyl-5-oxapentacyclo[9.7.0.02,8.04,6.012,16]octadecan-15-ol
Molecular Weight (g/mol) 304.467
Formula Weight 304.24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.