missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS EphB1 Functional Assay
Description
The PathHunter™ U2OS EphB1 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated EphB1 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged EphB1 and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Receptor Activation |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Recombinant Mouse Ephrin-B2Fc Chimera, CF |
| Quantity | 2 Vials |
| Target Entry | EphB1 |
| Accession Number | NM_004441.3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?