missing translation for 'onlineSavingsMsg'
Learn More

(2Alpha,3Beta)-2,3-Dihydroxy-urs-12-en-28-oic Acid, TRC

Product Code. 30137864
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137864 50 mg 50mg
30137134 100 mg 100mg
30003968 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137864 Supplier Toronto Research Chemicals Supplier No. TRCC69560050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4547-24-4
Molecular Formula C30 H48 O4
Molecular Weight (g/mol) 472.7
Synonym Urs-12-en-28-oic acid, 2,3-dihydroxy-, (2α,3β)-, Urs-12-en-28-oic acid, 2α,3β-dihydroxy- (7CI,8CI), (2α,3β)-2,3-Dihydroxyurs-12-en-28-oic acid, 2alpha-Hydroxyursolic acid, 2α,2β,-Dihydroxy-12-en-28-ursolic acid, 2α,3β-Dihydroxy ursolic acid, 2α,3β-Dihydroxylursolic acid, 2α,3β-Dihydroxyurs-12-en-28-oic acid, 2α-Hydroxyursolic acid, Colosic acid, Colosolic acid, Corosolic acid, Corsolic acid, Glucosol
IUPAC Name (1S,2R,4aS,6aR,6aS,6bR,8aR,10R,11R,12aR,14bS)-10,11-dihydroxy-1,2,6a,6b,9,9,12a-heptamethyl-2,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydro-1H-picene-4a-carboxylic acid
SMILES C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C30H48O4/c1-17-10-13-30(25(33)34)15-14-28(6)19(23(30)18(17)2)8-9-22-27(5)16-20(31)24(32)26(3,4)21(27)11-12-29(22,28)7/h8,17-18,20-24,31-32H,9-16H2,1-7H3,(H,33,34)/t17-,18+,20-,21+,22-,23+,24+,27+,28-,29-,30+/m1/s1
Formula Weight 472.3553
Chemical Name or Material Corosolic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.