missing translation for 'onlineSavingsMsg'
Learn More

2Alpha,3Alpha:16Alpha,17Alpha-Diepoxy-17Beta-acetoxy-5Alpha-androstane, TRC

Product Code. 30387065
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387065 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387065 Supplier Toronto Research Chemicals Supplier No. TRCD441895100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2alpha,3alpha:16alpha,17alpha-Diepoxy-17beta-acetoxy-5alpha-androstane
CAS 50588-22-2
Molecular Formula C21H30O4
Quantity 100 mg
Synonym 2alpha,3alpha:16alpha,17alpha-Bisepoxy-5alpha-androstan-17beta-ol acetate, 2alpha,3alpha:16alpha,17alpha-Diepoxy-17-acetoxy-5alpha-androstane
InChI Formula InChI=1S/C21H30O4/c1-11(22)24-21-18(25-21)9-15-13-5-4-12-8-16-17(23-16)10-19(12,2)14(13)6-7-20(15,21)3/h12-18H,4-10H2,1-3H3/t12-,13+,14-,15-,16-,17+,18+,19-,20-,21+/m0/s1
SMILES CC(=O)O[C@@]12O[C@@H]1C[C@H]3[C@@H]4CC[C@H]5C[C@@H]6O[C@@H]6C[C@]5(C)[C@H]4CC[C@]23C
Molecular Weight (g/mol) 346.46
Formula Weight 346.2144
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.