missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 2A3
GREENER_CHOICE

Product Code. 18728786
Click to view available options
:
10 mg
This item is not returnable. View return policy

Product Code. 18728786

Brand: Tocris Bioscience™ 7376/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent for RNA SHAPE-MaP experiments in vivo; cell permeable

2A3 is an electrophilic chemical probe that acylates the 2'-OH in the RNA backbone. 2A3 can be used for RNA SHAPE-MaP experiments in cell lines or live bacteria. It labels single-stranded or flexible regions more than folded regions (SHAPE methodology) and enables analysis of RNA secondary structures at single nucleotide resolution. For intracellular experiments, 2A3 has higher signal-to-noise ratio and prediction accuracy than NAI (Cat. No. 7004). 2A3 is cell permeable, and can probe both gram-positive and gram-negative bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2-Aminopyridin-3-yl)(1 H-imidazol-1-yl)methanone
Quantity 10 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C9H8N4O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.